missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog No. AAL1924903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1924903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1924903 Supplier Thermo Scientific Chemicals Supplier No. L1924903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3550-21-8 | Molecular Formula: C14H11NS | Molecular Weight: 225.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3550-21-8
Molecular Formula C14H11NS
Molecular Weight (g/mol) 225.31
MDL Number MFCD00046815
InChI Key WDOSFTZMBFYTED-UHFFFAOYSA-N
Synonym benzhydryl isothiocyanate, isothiocyanatodiphenylmethane, benzhydrylisothiocyanate, isothiocyanato phenyl methyl benzene, diphenylmethylisothiocyanate, benzene, 1,1'-isothiocyanatomethylene bis, diphenylmethanisothiocyanate, acmc-20alq6, benzhydryl-isothiocyanate, benzhydryl isothio-cyanate
PubChem CID 520742
IUPAC Name [isothiocyanato(phenyl)methyl]benzene
SMILES S=C=NC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Boiling Point 160°C to 165°C (4 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 2210613
Sensitivity Moisture sensitive
Formula Weight 225.31
Percent Purity 97%
Chemical Name or Material Benzhydryl isothiocyanate
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only