Learn More
Benzhydryl bromide, 90+%
CAS: 776-74-9 | C13H11Br | 247.135 g/mol
Supplier: Thermo Scientific Chemicals L0221136
Description
Benzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Protect from humidity and water. Store under dry inert gas.
Specifications
| Benzhydryl bromide | |
| 36°C to 41°C | |
| >110°C (230°F) | |
| C13H11Br | |
| 500 g | |
| 638544 | |
| bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane | |
| OQROAIRCEOBYJA-UHFFFAOYSA-N | |
| [bromo(phenyl)methyl]benzene | |
| 236603 | |
| ≥90% |
| 776-74-9 | |
| 183°C to 185°C (20 mmHg) | |
| Pungent | |
| MFCD00000134 | |
| UN1770 | |
| Moisture sensitive | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)Br | |
| 247.135 | |
| 247.14 |
Chemical Identifiers
| 776-74-9 | |
| 247.135 | |
| OQROAIRCEOBYJA-UHFFFAOYSA-N | |
| 236603 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)Br |
| C13H11Br | |
| MFCD00000134 | |
| bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane | |
| [bromo(phenyl)methyl]benzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIPHENYLMETHYL BROMIDE
EINECSNumber : 212-279-8
RTECSNumber : PA5350000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only