accessibility menu, dialog, popup

UserName

CAS RN 776-74-9

IUPAC Name: [bromo(phenyl)methyl]benzene
Synonyms: bromodiphenylmethane
Molecular Weight (g/mol): 247.14
Molecular Formula: C13H11Br
InChi Key: OQROAIRCEOBYJA-UHFFFAOYSA-N
SMILES: BrC(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)