missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl bromide, 90+%

CAS: 776-74-9 | C13H11Br | 247.135 g/mol

Supplier:  Thermo Scientific Chemicals L0221122

 View more versions of this product

Catalog No. AAL0221122


Only null left
Explore available promotions

Description

Description

Benzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Protect from humidity and water. Store under dry inert gas.
Specifications

Specifications

Benzhydryl bromide
36°C to 41°C
>110°C (230°F)
C13H11Br
100 g
638544
bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
OQROAIRCEOBYJA-UHFFFAOYSA-N
[bromo(phenyl)methyl]benzene
236603
≥90%
776-74-9
183°C to 185°C (20 mmHg)
Pungent
MFCD00000134
UN1770
Moisture sensitive
Insoluble in water.
C1=CC=C(C=C1)C(C2=CC=CC=C2)Br
247.135
247.14

Chemical Identifiers

776-74-9
247.135
OQROAIRCEOBYJA-UHFFFAOYSA-N
236603
C1=CC=C(C=C1)C(C2=CC=CC=C2)Br
C13H11Br
MFCD00000134
bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
[bromo(phenyl)methyl]benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIPHENYLMETHYL BROMIDE

EINECSNumber : 212-279-8

RTECSNumber : PA5350000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only