missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, 0.004 M (N), Ricca Chemical

Catalog No. 95516
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Cubitainer
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
95516 500 mL Poly Bottle
9555 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 95516 Supplier Ricca Chemical Company Supplier No. 95516

Chemical Identifiers

CAS 121-54-0 , 7732-18-5
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.088
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

Color Colorless
CAS Min % 98.0, 0.18
CAS Max % 100.0, 0.18
Grade Laboratory
Packaging Poly Bottle
pH 5.5 to 7.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.004Molar
Chemical Name or Material Benzethonium Chloride