missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl hydrazide, 98%

CAS: 80-17-1 | C6H8N2O2S | 172.20 g/mol

Supplier:  Thermo Scientific Chemicals B2397122

 View more versions of this product

Catalog No. AAB2397122


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzenesulfonyl hydrazide
101°C to 110°C (decomposition)
MFCD00007583,MFCD08064450
UN3226
14,7598
VJRITMATACIYAF-UHFFFAOYSA-N
benzenesulfonohydrazide
65723
98%
80-17-1
C6H8N2O2S
100 g
640079
benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
NNS(=O)(=O)C1=CC=CC=C1
172.20
172.21

Chemical Identifiers

80-17-1
172.20
VJRITMATACIYAF-UHFFFAOYSA-N
65723
NNS(=O)(=O)C1=CC=CC=C1
C6H8N2O2S
MFCD00007583,MFCD08064450
benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
benzenesulfonohydrazide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H242-H228-H301
Heating may cause a fire.
Flammable solid.
Toxic if swallowed.

P210-P220-P234-P240-P241-P280-P370+P378q-P375-P380-P501c

H228-H242

DOTInformation : Hazard Class: 4.1

EINECSNumber : 201-255-2

RTECSNumber : DB6888000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only