missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl hydrazide, 98%

Catalog No. p-7027310
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2397114 25 g
AAB2397122 100 g
AAB2397136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2397114 Supplier Thermo Scientific Chemicals Supplier No. B2397114
Only null left

CAS: 80-17-1 | C6H8N2O2S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-17-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00007583,MFCD08064450
InChI Key VJRITMATACIYAF-UHFFFAOYSA-N
Synonym benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
PubChem CID 65723
IUPAC Name benzenesulfonohydrazide
SMILES NNS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 101°C to 110°C (decomposition)
Quantity 25 g
UN Number UN3226
Beilstein 640079
Merck Index 14,7598
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl hydrazide
Hazard Category H228-H242
Hazard Statement GHS H Statement
H242-H228-H301
Heating may cause a fire.
Flammable solid.
Toxic if swallowed.
Precautionary Statement P210-P220-P234-P240-P241-P280-P370+P378q-P375-P380-P501c
DOTInformation Hazard Class: 4.1
EINECSNumber 201-255-2
RTECSNumber DB6888000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only