missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 3562-84-3
Molecular Formula C17H12Br2O3
Molecular Weight (g/mol) 424.08
InChI Key WHQCHUCQKNIQEC-UHFFFAOYSA-N
Synonym benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
PubChem CID 2333
ChEBI CHEBI:3023
IUPAC Name (3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
SMILES CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC460900010
View Documents
Thermo Scientific Chemicals
460900010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC460900050
View Documents
Thermo Scientific Chemicals
460900050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3562-84-3
424.08
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
CHEBI:3023
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
C17H12Br2O3
WHQCHUCQKNIQEC-UHFFFAOYSA-N
2333
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone

Specifications

3562-84-3
Authentic
Glass Bottle
1g
Solubility in water: .. Other solubilities: soluble in hot methanol
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
424.08
CHEBI:3023
98%
0.5% max.
97.5% min. (GC)
C17H12Br2O3
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
WHQCHUCQKNIQEC-UHFFFAOYSA-N
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
2333
424.08
Benzbromarone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 222-630-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only