accessibility menu, dialog, popup

UserName

H3COOBrHOBr

CAS RN 3562-84-3

IUPAC Name: 2,6-dibromo-4-(2-ethyl-1-benzofuran-3-carbonyl)phenol
Synonyms: benzbromarone normurat AL, benzbromaron benzbromaron ratiopharm urinorm exurate desuric aliud brand of benzbromarone benzbromarone heumann brand benzbromaronum
Molecular Weight (g/mol): 424.09
Molecular Formula: C17H12Br2O3
InChi Key: WHQCHUCQKNIQEC-UHFFFAOYSA-N
SMILES: CCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)