missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzanthrone 98.0+%, TCI America™
$87.60 - $726.98
Chemical Identifiers
| CAS | 82-05-3 |
|---|---|
| Molecular Formula | C17H10O |
| Molecular Weight (g/mol) | 230.266 |
| MDL Number | MFCD00003585 |
| InChI Key | HUKPVYBUJRAUAG-UHFFFAOYSA-N |
| Synonym | benzanthrone, 7h-benz de anthracen-7-one, benzanthrenone, ms-benzanthrone, dye, benzanthrone, benzoanthrone, mesobenzanthrone, 7h-benzo de anthracen-7-one, 1,9-benzanthrone |
| PubChem CID | 6697 |
| IUPAC Name | benzo[a]phenalen-7-one |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O |
Chemical Identifiers
| 82-05-3 | |
| 230.266 | |
| HUKPVYBUJRAUAG-UHFFFAOYSA-N | |
| 6697 | |
| C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O |
| C17H10O | |
| MFCD00003585 | |
| benzanthrone, 7h-benz de anthracen-7-one, benzanthrenone, ms-benzanthrone, dye, benzanthrone, benzoanthrone, mesobenzanthrone, 7h-benzo de anthracen-7-one, 1,9-benzanthrone | |
| benzo[a]phenalen-7-one |
Specifications
| 82-05-3 | |
| C17H10O | |
| 25 g | |
| HUKPVYBUJRAUAG-UHFFFAOYSA-N | |
| benzo[a]phenalen-7-one | |
| 6697 | |
| ≥98.0% (GC) | |
| Benzanthrone |
| 176°C | |
| MFCD00003585 | |
| benzanthrone, 7h-benz de anthracen-7-one, benzanthrenone, ms-benzanthrone, dye, benzanthrone, benzoanthrone, mesobenzanthrone, 7h-benzo de anthracen-7-one, 1,9-benzanthrone | |
| C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O | |
| 230.266 | |
| 230.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0793
RTECSNumber : CX5075000
TSCA : Yes