CAS RN 82-05-3
IUPAC Name:
tetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,9,11,13,15-octaen-8-one
Synonyms:
benzanthrone
Molecular Weight (g/mol):
230.27
Molecular Formula:
C17H10O
InChi Key:
HUKPVYBUJRAUAG-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2C2=C3C(C=CC=C13)=CC=C2
1
–
1
of
1
results
1
Benzanthrone 98.0+%, TCI America™
CAS: 82-05-3 Molecular Formula: C17H10O Molecular Weight (g/mol): 230.266 MDL Number: MFCD00003585 InChI Key: HUKPVYBUJRAUAG-UHFFFAOYSA-N Synonym: benzanthrone,7h-benz de anthracen-7-one,benzanthrenone,ms-benzanthrone,dye, benzanthrone,benzoanthrone,mesobenzanthrone,7h-benzo de anthracen-7-one,1,9-benzanthrone PubChem CID: 6697 IUPAC Name: benzo[a]phenalen-7-one SMILES: C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O