missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol

$348.63 - $562.92

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC263820010
SDS View Documents
Thermo Scientific Chemicals
263820010
1 L Glass bottle
Each for $348.63
Only null left
 
AC263820025
SDS View Documents
Thermo Scientific Chemicals
263820025
2.5 L Glass bottle
Each for $562.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8001-54-5
170.66
Alkyl-benzyl-dimethylammonium chloride
C9H13ClN
MFCD00145757

Specifications

Colorless to Yellow
>100.0°C
52.0,51.0
C6H5CH2N(CH3)2RClR=C8H17toC18H37
49 to 52% (based on M.W.= 352.5)
15, 1061
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether
Glass bottle
130 mPa.s (20°C)
1 L
Solution
8001-54-5,7732-18-5
49.0,48.0
C9H13ClN
MFCD00145757
0.9800g/mL
Alkyl-benzyl-dimethylammonium chloride
170.66
0.98
6 to 9 (10% aq. soln.)
105°C
Benzalkonium chloride
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed. Very toxic to aquatic life with long lasting effects.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only