missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalkonium Chloride, 17% (w/v), Solution, Spectrum™ Chemical
B1069, 8001-54-5
$431.06 - $22748.76
Chemical Identifiers
| CAS | 8001-54-5 |
|---|---|
| Molecular Formula | C9H13ClN |
| Molecular Weight (g/mol) | 170.66 |
| SMILES | [Cl-].C[N+](C)(*)CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600879
|
Spectrum Chemical Mfg Cor
B1069500MLGL |
500 mL | Amber Glass Bottle |
Each for $431.06
|
|
||||
|
18600880
|
Spectrum Chemical Mfg Cor
B10694LTGL |
4 L | Amber Glass Bottle |
Each for $2,853.97
|
|
||||
|
18600881
|
Spectrum Chemical Mfg Cor
B1069200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Benzalkonium Chloride, 17 Percent (w/v), Solution is also referred to as BZK is an aqueous solution for handling purposes. It varies in color, from colorless to a pale yellow and has an almond-like smell and bitter taste. Its uses vary from disinfectant to microbial corrosion inhibition and can also be used as an algaecide.Chemical Identifiers
| 8001-54-5 | |
| C9H13ClN | |
| 170.66 | |
| [Cl-].C[N+](C)(*)CC1=CC=CC=C1 |
Specifications
| 8001-54-5,7732-18-5 | |
| Amber Glass Bottle | |
| 500 mL | |
| [Cl-].C[N+](C)(*)CC1=CC=CC=C1 | |
| 17% ±1.5 |
| 17%,83% | |
| C9H13ClN | |
| UN1760 | |
| 170.66 | |
| Chemical Solution |