missing translation for 'onlineSavingsMsg'
Learn More

Benz[a]anthracene 98.0+%, TCI America™

Catalog No. p-7118303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00171G 1 g
B00175G 5 g
B001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B00171G Supplier TCI America Supplier No. B00171G
Only null left

Chemical Identifiers

CAS 56-55-3
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
MDL Number MFCD00003599
InChI Key DXBHBZVCASKNBY-UHFFFAOYSA-N
Synonym benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene
PubChem CID 5954
ChEBI CHEBI:51348
IUPAC Name tetraphene
SMILES C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1

Specifications

Melting Point 161°C
Boiling Point 438°C
Quantity 1 g
UN Number 3077
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benz[a]anthracene
RTECSNumber CV9275000
TSCA Yes