missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benedict's Reagent, Qualitative, for detection of reducing sugars, Ricca Chemical
Detection of reducing sugars
$80.03 - $368.44
Specifications
| CAS | 7732-18-5,497-19-8,7758-99-8 |
|---|---|
| CAS Min % | 76.0,13.15,7.6,1.31 |
| CAS Max % | 79.0,13.68,7.91,1.37 |
| Grade | Laboratory |
| pH | ∼10.6 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
930-4
|
Ricca Chemical Company
RI9304 |
120 mL | Natural Poly Bottle |
Each for $80.03
|
|
||||
|
930-16
|
Ricca Chemical Company
93016 |
500 mL | Natural Poly Bottle |
Each for $84.24
|
|
||||
|
930-32
|
Ricca Chemical Company
93032 |
1 L | Natural Poly Bottle |
Each for $129.66
|
|
||||
|
930-1
|
Ricca Chemical Company
RI9301 |
4 L | Natural Poly Bottle |
Each for $368.44
|
|
||||
Specifications
| 7732-18-5,497-19-8,7758-99-8 | |
| 79.0,13.68,7.91,1.37 | |
| ∼10.6 |
| 76.0,13.15,7.6,1.31 | |
| Laboratory |