missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benedict's Reagent, Qualitative, for detection of reducing sugars, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Benedict's Reagent |
| Color | Blue |
| Name Note | Qualitative |
| CAS | 7732-18-5, 63126-89-6, 497-19-8, 7758-99-8 |
| CAS Min % | 76.0, 13.15, 7.6, 1.31 |
| CAS Max % | 79.0, 13.68, 7.91, 1.37 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | ∼10.6 |
| Quantity | 120 mL |
| Show More |