Learn More
Benedict Qualitative Solution, Certified, LabChem™
Supplier: LabChem LC116504
Specifications
| Benedict Qualitative Solution | |
| 70.97,17.3,10,1.73 | |
| CuSO4·5H2O | |
| 249.68 | |
| Passes Test | |
| Fume; Carbon monoxide; Carbon dioxide | |
| Passes Test |
| Blue | |
| 63126-89-6 , 497-19-8 , 7758-99-8 | |
| Soluble in water | |
| Certified | |
| Poly Bottle | |
| 4 L | |
| Liquid |