missing translation for 'onlineSavingsMsg'
Learn More

Belleau's Reagent [Sulfurating Reagent], TCI America™

Catalog No. B15175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15175G Supplier TCI America Supplier No. B15175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88816-02-8
Molecular Formula C24H18O2P2S4
Molecular Weight (g/mol) 528.594
MDL Number MFCD00191383
InChI Key BFIWQSSAMKDRRZ-UHFFFAOYSA-N
Synonym 2,4-Bis(4-phenoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulfide
PubChem CID 11813414
IUPAC Name 2,4-bis(4-phenoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)P3(=S)SP(=S)(S3)C4=CC=C(C=C4)OC5=CC=CC=C5

Specifications

Melting Point 197°C
Color Yellow
Quantity 5 g
Formula Weight 528.59
Physical Form Crystalline Powder
Chemical Name or Material Belleau′s Reagent [Sulfurating Reagent]