missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks, Holds Tubes 10 to 13 mm
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
$218.74 - $240.72
Specifications
| Product Type | Test Tube Rack |
|---|---|
| Autoclavable | Autoclavable |
| Holds | Tubes (10 to 13 mm) |
| Material | Epoxy-coated Steel |
| Shape | Rectangular |
| Catalog Number | Mfr. No. | Color | Positions | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Positions | Price | Quantity | ||||
|
22-260541
|
Bel-Art™
F187500001 |
Blue | 72 |
Each for $237.34
Case of 24 Each for $5,696.16
|
|
||||
|
22-260545
|
Bel-Art™
F187530001 |
Blue | 108 |
Each for $240.72
Case of 24 Each for $5,777.28
|
|
||||
|
14-793-231
|
N/A | Blue | 60 |
Each for $218.74
Case of 24 Each for $5,249.76
|
|
||||
|
14-793-235
|
N/A | Blue | 150 |
Each for $219.48
Case of 12 Each for $2,633.76
|
|
||||
Description
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
| Test Tube Rack | |
| Tubes (10 to 13 mm) | |
| Rectangular | |
| 13 mm |
| Autoclavable | |
| Epoxy-coated Steel | |
| Non-sterile |