missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Poxygrid™ Wire Test Tube Racks, Holds Tubes 10 to 13 mm

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

 View more versions of this product

Catalog No. 14-793-231



Only null left
Explore available promotions

Description

Description

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (10 to 13 mm)
60
3 Tiers
Non-sterile
17.5 cm
11.4 cm
6.4 cm
60 mm
6 x 10
Blue
Epoxy-coated Steel
Rectangular
Yes, When Empty
6.9 x 4.5 x 2.5 in. (17.5 x 11.4 x 6.4cm)
6.9 in.
4.5 in.
2.5 in.
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions