missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks, Holds Tubes 10 to 13 mm
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Supplier: Bel-Art™ F187530001
Description
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
| Test Tube Rack | |
| Autoclavable | |
| Tubes (10 to 13 mm) | |
| 108 | |
| 3 Tiers | |
| Non-sterile | |
| 20.5 cm | |
| 16.4 cm | |
| 6.5 cm | |
| 13 mm |
| 9 x 12 | |
| Blue | |
| Epoxy-coated Steel | |
| Rectangular | |
| Yes, When Empty | |
| 8 x 6.5 x 2.5 in. (20.5 x 16.4 x 6.4cm) | |
| 8.07 in. | |
| 6.45 in. | |
| 2.51 in. |