Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Mini Sampler Spoons

Avoid cross contamination by using a different spoon for each sample

$52.77 - $532.00

Specifications

Color White
Product Type Sample Spoon
Disposable Yes
For Use With (Application) Designed for retrieving small, specific volume samples of powders or similar materials
Material Polystyrene
View More Specs
Products
Catalog Number Mfr. No. Capacity Price Quantity    

03-410-518

 
bel-art™
F367210010
0.0034 oz., 0.1mL Pack of 25 Each for $56.23
Case of 10 Pack for $562.30

03-410-517

 
bel-art™
F367210005
0.0016 oz., 0.05mL Pack of 25 Each for $64.50
Case of 10 Pack for $645.00

03-410-520

 
bel-art™
F367210050
0.017 oz., 0.5mL Pack of 25 Each for $52.77
Case of 10 Pack for $527.70

03-410-519

 
bel-art™
F367210025
0.0085 oz., 0.25mL Pack of 25 Each for $56.23
Case of 10 Pack for $562.30

03-410-521

 
bel-art™
367210100
0.034 oz., 1mL Case of 250 Each for $532.00
Description & Specifications

Specifications

Color White
Product Type Sample Spoon
Disposable Yes
For Use With (Application) Designed for retrieving small, specific volume samples of powders or similar materials
Material Polystyrene
Sterile Yes

  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.