Learn More
Bel-Art™ Mini Sampler Spoons
Avoid cross contamination by using a different spoon for each sample
Supplier: Bel-Art™ F367210005
Description
- These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
- Made from white high-impact polystyrene plastic
- All sterile products are lot stamped for accurate tracing
Specifications
| Sterile | |
| 0.05 mL | |
| Single-use | |
| Designed for retrieving small, specific volume samples of powders or similar materials |
| Not Autoclavable | |
| White | |
| Spoon | |
| Polystyrene |