Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
$149.80 - $238.34
Specifications
Autoclavable Autoclavable
Material Polypropylene
Includes Reducing connector
Color Clear
View More Specs
Products 5
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
22235697
View Documents Promotion Details
Bel-Art™
H195580000
4.7 to 6mm
Pack of 12 Each for $149.80
Only null left
22235705
View Documents Promotion Details
Bel-Art™
H195590000
4.7 to 9.5mm
Pack of 12 Each for $238.34
Only null left
22235721
View Documents Promotion Details
Bel-Art™
H195610000
Fits 9 x 6mm Tubing I.D.
Pack of 12 Each for $154.97
Only null left
22235739
View Documents Promotion Details
Bel-Art™
H195620000
6 to 12.7mm
Pack of 12 Each for $166.44
Only null left
22235747
View Documents Promotion Details
Bel-Art™
H195630000
9.5 to 12.7mm
Pack of 12 Each for $149.80
Only null left
Description
Description
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
Autoclavable
Polypropylene
Reducing connector
Clear