Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
Supplier: Bel-Art™ H195580000
Description
- Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Chemical Identifiers
| Autoclavable | |
| Polypropylene | |
| Reducing connector | |
| Clear |
Specifications
| Autoclavable | |
| 4.7 to 6mm | |
| Reducing connector | |
| Barbed Fitting |
| Polypropylene | |
| For 3/16 in. to 1/4 in. tubing (4.7 to 6mm) | |
| Clear |