missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Antibody Saver Trays
Minimize the use of expensive antibodies during incubations by using an optimally sized tray
$117.90 - $402.79
Specifications
| Color | Natural |
|---|---|
| Height (Metric) | 25 mm |
| Material | Polypropylene |
| Type | Collection Tray |
| Includes | Attached lid to protect against spillage |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|
03421133
|
Bel Art™
F451000000 |
0.62 x 4.5 in. |
Pack of 5 Each for $117.90
Case of 6 Pack for $707.40
|
|
|||||
|
03421135
|
Bel Art™
F451000002 |
6.5 x 6.5 in. |
Pack of 5 Each for $402.79
Case of 6 Pack for $2,416.74
|
|
|||||
|
03421134
|
Bel Art™
F451000001 |
3.5 x 3.5 in. (9 x 9 cm) |
Pack of 5 Each for $150.69
Case of 6 Pack for $904.14
|
|
|||||
Description
- For popular midi and mini gels
- Stackable, reclosable and reusable
- All trays include attached lid to protect against spillage
- Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
- Polypropylene; autoclavable
Specifications
| Natural | |
| Polypropylene | |
| Attached lid to protect against spillage |
| 25 mm | |
| Collection Tray | |
| Autoclavable |