missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Antibody Saver Trays
Minimize the use of expensive antibodies during incubations by using an optimally sized tray
Supplier: Bel Art™ F451000002
Description
- For popular midi and mini gels
- Stackable, reclosable and reusable
- All trays include attached lid to protect against spillage
- Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
- Polypropylene; autoclavable
Chemical Identifiers
| Natural | |
| Polypropylene | |
| Attached lid to protect against spillage |
| 25 mm | |
| Collection Tray | |
| Autoclavable |
Specifications
| 680 cm3 | |
| Natural | |
| 16.5 cm | |
| Collection Tray | |
| Attached lid to protect against spillage | |
| Square | |
| Autoclavable |
| 6.5 x 6.5 in. | |
| 25 mm | |
| Polypropylene | |
| 16.5 cm | |
| 6.5 in. | |
| 6.5 in. |