missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
$603.61 - $1,930.71
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact |
| Closure Type | Zip Tie |
| Color | Red |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity & Availability | |||||
|
03411700
|
Bel-Art™
F131641419 |
2 to 4 gal. |
Pack of 200 Each for $603.61
|
|
|||||
|
03411701
|
Bel-Art™
F131641923 |
5 to 9 gal. |
Pack of 200 Each for $824.82
|
|
|||||
|
03411702
|
Bel-Art™
F131642535 |
13 to 20 gal. |
Pack of 200 Each for $1,461.68
|
|
|||||
|
03411703
|
Bel-Art™
F131643138 |
20 to 30 gal. |
Pack of 200 Each for $1,930.71
|
|
|||||
Description
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
- Bright red polypropylene bags include black biohazard symbol
- Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135°C (275°F)
Specifications
| Autoclavable | |
| Zip Tie | |
| Single-use | |
| Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
| Non-sterile | |
| Biohazard Disposal Bag |
| ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
| Red | |
| Used for biohazard waste | |
| Polypropylene | |
| 1.50 mil | |
| 0.279 mm |