missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
Supplier: Bel-Art™ F131641923
Description
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
- Bright red polypropylene bags include black biohazard symbol
- Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135°C (275°F)
Chemical Identifiers
| Autoclavable | |
| Zip Tie | |
| Single-use | |
| Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
| Non-sterile | |
| Biohazard Disposal Bag |
| ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
| Red | |
| Used for biohazard waste | |
| Polypropylene | |
| 1.50 mil | |
| 0.279 mm |
Specifications
| 19W x 23H in. (48 x 58cm) | |
| ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
| Red | |
| Used for biohazard waste | |
| Polypropylene | |
| 1.50 mil | |
| 23 in. | |
| 23 in. | |
| Biohazard Disposal Bag | |
| 26 L | |
| 48 cm |
| Autoclavable | |
| Zip Tie | |
| Single-use | |
| Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
| Non-sterile | |
| 19 x 23 in. | |
| 19 in. | |
| 58 cm | |
| 5 to 9 gal. | |
| 58 cm | |
| 0.279 mm |