missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil

Catalog No. 03411703
Change view
Click to view available options
Capacity (English):
13 to 20 gal.
2 to 4 gal.
20 to 30 gal.
5 to 9 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
03411703 20 to 30 gal.
03411700 2 to 4 gal.
03411701 5 to 9 gal.
03411702 13 to 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03411703 Supplier Bel-Art™ Supplier No. F131643138

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.

  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)

Specifications

Description 31W x 38H in. (79 x 97cm)
Autoclavable Autoclavable
Certifications/Compliance ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) Used for biohazard waste
Includes Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
Material Polypropylene
Sterility Non-sterile
Thickness (English) 1.50 mil
Dimensions (L x W) 31 x 38 in.
Length (English) 38 in.
Width (English) 31 in.
Height (English) 38 in.
Height (Metric) 97 cm
Product Type Biohazard Disposal Bag
Capacity (English) 20 to 30 gal.
Capacity (Metric) 95 L
Length (Metric) 97 cm
Width (Metric) 79 cm
Thickness (Metric) 0.279 mm
Show More Show Less