missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone dipropionate, ≥99%, MilliporeSigma™ Supelco™

Beclomethasone dipropionate is a type of steroid drug, administered by means of inhalation. It can behave like an anti-inflammatory agent within the respiratory tract. It can also decrease airway resistance and reduce mucosal oedema.

$163.90 - $2053.63

Specifications

CAS 9-8-5534
Physical Form Neat
Linear Formula C28H37 ClO7
MDL Number MFCD00135613
Synonym 9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012214
View Documents
Merck Emd Millipore
B3022250MG
250 mg
Each for $163.90
Only null left
 
111012212
View Documents
Merck Emd Millipore
SGMAB30221G
1 g
Each for $556.10
Only null left
 
111012213
View Documents
Merck Emd Millipore
B30225G
5 g
Each for $2,053.63
Only null left
 
Description

Description

Specifications

Specifications

9-8-5534
C28H37 ClO7
9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
Drug Analysis
≥99%
Neat
MFCD00135613
Analytical Standard
521.04
Analytical Standard
Videos
SDS
Documents

Documents