missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclomethasone dipropionate, ≥99%, MilliporeSigma™ Supelco™
Beclomethasone dipropionate is a type of steroid drug, administered by means of inhalation. It can behave like an anti-inflammatory agent within the respiratory tract. It can also decrease airway resistance and reduce mucosal oedema.
Supplier: Merck Emd Millipore SGMAB30221G
Specifications
| 9-8-5534 | |
| C28H37 ClO7 | |
| 9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate | |
| Drug Analysis | |
| 521.04 | |
| Analytical Standard |
| Neat | |
| MFCD00135613 | |
| Analytical Standard | |
| 1 g | |
| ≥99% |