missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD140 98.0+%, TCI America™
$533.13 - $1535.98
Chemical Identifiers
| CAS | 1201643-08-4 |
|---|---|
| Molecular Formula | C21H21BF2N2O |
| Molecular Weight (g/mol) | 366.219 |
| InChI Key | NDQRWYKJLFZGBK-RMKNXTFCSA-N |
| Synonym | 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene |
| PubChem CID | 72198885 |
| SMILES | [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D489825MG
|
TCI America
D489825MG |
25 mg |
Each for $533.13
|
|
|||||
|
D4898100MG
|
TCI America
D4898100MG |
100 mg |
Each for $1,535.98
|
|
|||||
Chemical Identifiers
| 1201643-08-4 | |
| 366.219 | |
| 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene | |
| [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F |
| C21H21BF2N2O | |
| NDQRWYKJLFZGBK-RMKNXTFCSA-N | |
| 72198885 |
Specifications
| 1201643-08-4 | |
| C21H21BF2N2O | |
| 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene | |
| [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F | |
| 72198885 | |
| ≥98.0% (GC) | |
| BD140 [for Albumin binding assay] |
| 155°C | |
| 25 mg | |
| NDQRWYKJLFZGBK-RMKNXTFCSA-N | |
| 366.219 | |
| 366.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No