missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| BD140 [for Albumin binding assay] | |
| 155°C | |
| 100 mg | |
| NDQRWYKJLFZGBK-RMKNXTFCSA-N | |
| 366.219 | |
| 366.22 | |
| Crystalline Powder |
| 1201643-08-4 | |
| C21H21BF2N2O | |
| 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene | |
| [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F | |
| 72198885 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1201643-08-4 | |
| 366.219 | |
| 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene | |
| [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F |
| C21H21BF2N2O | |
| NDQRWYKJLFZGBK-RMKNXTFCSA-N | |
| 72198885 |
Safety and Handling
TSCA : No