missing translation for 'onlineSavingsMsg'
Learn More

BD Falcon™ RODAC™ Plate

Catalog No. 08757152
Change view
Click to view available options
Diameter (Metric):
65 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
08757152 65 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08757152 Supplier BD Supplier No. 210340
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for environmental studies of microbial contamination

  • 10mm grid with 21 squares
  • Surface area: approximately 25cm2
  • Sterile, with lid
  • Disposable

Order Info

20 per sleeve.

Specifications

Closure Included Yes
Culture Area ∼25 cm2
Disposable Single-use
Sterility Sterile
Packaging Sleeve
Quantity 20/Sleeve
Height (Metric) 15 mm
Diameter (Metric) 65 mm