missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ TC Yeastolate, ultra-filtered (UF)

Enhance cell culture performance with Gibco™ Difco™ TC Yeastolate Ultra-Filtered (UF) Low Bioburden while also mitigating risk in bioprocessing methods.

$517.96 - $54353.25

Specifications

Color Beige
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type TC Yeastolate
For Use With (Application) Preparing bacterial, insect, and mammalian cell culture media
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Manufacturing Quality Price Quantity  
Catalog Number Mfr. No. Quantity Manufacturing Quality Price Quantity  
B92804
View Documents
Gibco™
292804
500 g UF
Each for $517.96
Only null left
 
B92805
View Documents
Gibco™
292805
10 kg UF
Each for $8,789.54
Only null left
 
670079
View Documents
Gibco™
670079
50 kg UF N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.

Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.

Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU

Specifications

Specifications

Beige
Powder
Preparing bacterial, insect, and mammalian cell culture media
Mammalian, Insect, Microbial
TC Yeastolate
Videos
SDS
Documents

Documents