Learn More
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
Supplier: BD 264010
Alert:
Not intended for human or therapeutic use.
Description
- Adjuvants are mixed with aqueous antigen solution or suspension
- Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
- With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
- Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
| Store in cool, dry conditions in well sealed containers, away from oxidizing agents. | |
| Immunological studies with laboratory animals | |
| 6 x 100 mg |
| Mycobacterium butyricum | |
| Killed M. butyricum for use in adjuvants |