missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Adjuvants
Description
- Adjuvants are mixed with aqueous antigen solution or suspension
- Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
- With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
- Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
Specifications
| Formulation | Suspension of 5mg M. butyricum in a mixture of paraffin oil and an emulsifying agent |
| Product Type | Freund's Complete Adjuvant |
| For Use With (Application) | Immunological studies with laboratory animals |
| Quantity | 6 x 10 mL |
| Content And Storage | Store in cool, dry conditions in well sealed containers, away from oxidizing agents. |