missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Yeast Extract

Water-soluble extract of autolyzed yeast cells suitable for use in culture media

Supplier:  Gibco™ 211930

 View more versions of this product

Catalog No. B11930

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

LUO, MUO
Bacteria
Powder
Pail
5 lb.
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Mammalian, Insect, Microbial
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
Animal origin-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions