missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptone

An animal origin (AO) pancreatic digest of casein.

Supplier:  Gibco™ 211699

 View more versions of this product

Catalog No. DF0123-07-5


Only null left
Explore available promotions

Description

Description

  • Bacto Tryptone is an animal origin (AO) pancreatic digest of casein
  • It is included in standard methods manual applications and is listed in the Reagent section of the United States Pharmacopeia as meeting the specifications for a pancreatic digest of casein, a component in many of the media listed
  • The nutritional profile of this supplement is unique in that it provides a range of nutrients
  • Customers have used this supplement in fermentation applications and shown that it successfully supports the growth of many bacterial species during the production of animal health vaccines
  • This includes Escherichia coli, and Fusobacterium and Agrobacterium species when used together with Bacto Yeast Extract
  • Store in cool, dry conditions in well-sealed containers at 2° to 25°C

Features:

  • Pancreatic digest of casein
  • Developed while investigating a peptone particularly suitable for the elaboration of indole by bacteria
  • Animal origin nutritional supplement

Applications of Bacto Tryptone include:

  • Works well in fermentation applications
  • Use with common organisms such as Escherichia coli, as well as uncommon organisms such as the diatom Nitzschia laevis
Specifications

Specifications

LUO, MUO
Bacterial Media
Dehydrated culture media
2 kg
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Pail
Bacto
Animal Origin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions