Learn More
Brady™ BBP33 Topcoated Polyimide Labels
Mark wires and cables, particularly in aerospace defense and mass transit settings. Brady™ BBP33 Topcoated Polyimide Labels are self extinguishing labels with very high temperature ratings.
Supplier:  Brady™ B33C3000472
 View more versions of this product
Catalog No. 19107897
Only null left
Description
Description
  • Can withstand very high temperatures
  • RoHS compliant
  • Used for wire and cable marking, particularly in aerospace defense and mass transit industries
Specifications
Chemical Identifiers
RoHS
Acrylic, Permanent
Cartridge
BBP™33 Printer
85 ft.
Chemical and humidity resistant
Matte
Polyimide (B-472)
0.0032 in.
Specifications
RoHS
Acrylic, Permanent
Matte
Polyimide (B-472)
BBP™33 Printer
85 ft.
Chemical and humidity resistant
White
Cartridge
Topcoated Polyimide Label
0.0032 in.
3.2 in.
Product Suggestions
Product Suggestions