missing translation for 'onlineSavingsMsg'
Learn More

BAY 11-7082 98.0+%, TCI America™

Catalog No. T2846100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2846100MG 100 mg
T28461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2846100MG Supplier TCI America Supplier No. T2846100MG

Chemical Identifiers

CAS 19542-67-7
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.25
MDL Number MFCD00712162
InChI Key DOEWDSDBFRHVAP-KRXBUXKQSA-N
Synonym e-3-tosylacrylonitrile, e-3-p-toluenesulfonyl acrylonitrile, unii-4y5g2a4f6o, e-3-4-methylphenyl sulfonylprop-2-enenitrile, 3-p-toluenesulfonyl acrylonitrile, e, 3-4-methylphenylsulfonyl-2-propenenitrile, e-3-4-methylphenylsulfonyl-2-propenenitrile, e 3-4-methylphenyl sulfonyl-2-propenenitrile, 2e-3-4-methylphenyl sulfonyl prop-2-enenitrile, 2-propenenitrile, 3-4-methylphenyl sulfonyl-, 2e
PubChem CID 5353431
ChEBI CHEBI:85928
IUPAC Name (2E)-3-(4-methylbenzenesulfonyl)prop-2-enenitrile
SMILES CC1=CC=C(C=C1)S(=O)(=O)\C=C\C#N

Specifications

Melting Point 134°C
Color White
Quantity 100 mg
Formula Weight 207.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material BAY 11-7082
RTECSNumber UD1430000
TSCA No
Recommended Storage Refrigerator