accessibility menu, dialog, popup

UserName

CAS RN 19542-67-7

IUPAC Name: (2E)-3-(4-methylbenzenesulfonyl)prop-2-enenitrile
Synonyms: (2E)-3-(4-methylbenzenesulfonyl)prop-2-enenitrile
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: DOEWDSDBFRHVAP-KRXBUXKQSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)\C=C\C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results