Learn More
Barium tartrate, 98%
CAS: 5908-81-6 | C4H4BaO6 | 285.397 g/mol
$40.44 - $117.75
Chemical Identifiers
| CAS | 5908-81-6 |
|---|---|
| Molecular Formula | C4H4BaO6 |
| Molecular Weight (g/mol) | 285.397 |
| MDL Number | MFCD00054437 |
| InChI Key | HQYOWPDUMCBSLQ-UHFFFAOYSA-L |
| Synonym | barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate |
| PubChem CID | 6453753 |
| IUPAC Name | barium(2+);2,3-dihydroxybutanedioate |
| SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1960418
|
Thermo Scientific Chemicals
A1960418 |
50 g |
Each for $40.44
|
|
|||||
|
AAA1960430
|
Thermo Scientific Chemicals
A1960430 |
250 g |
Each for $117.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5908-81-6 | |
| 285.397 | |
| HQYOWPDUMCBSLQ-UHFFFAOYSA-L | |
| 6453753 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] |
| C4H4BaO6 | |
| MFCD00054437 | |
| barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate | |
| barium(2+);2,3-dihydroxybutanedioate |
Specifications
| 5908-81-6 | |
| C4H4BaO6 | |
| UN1564 | |
| barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate | |
| HQYOWPDUMCBSLQ-UHFFFAOYSA-L | |
| barium(2+);2,3-dihydroxybutanedioate | |
| 6453753 | |
| 98% | |
| 2.98 |
| 50 g | |
| MFCD00054437 | |
| 6120238 | |
| Very slightly soluble in water | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] | |
| 285.397 | |
| 285.41 | |
| Odorless | |
| Barium tartrate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 227-615-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only