missing translation for 'onlineSavingsMsg'
Learn More

Barium tartrate, 98%

CAS: 5908-81-6 | C4H4BaO6 | 285.397 g/mol

Supplier:  Thermo Scientific Chemicals A1960430

 View more versions of this product

Catalog No. AAA1960430


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Barium tartrate
250 g
MFCD00054437
6120238
Very slightly soluble in water
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2]
285.397
285.41
Odorless
5908-81-6
C4H4BaO6
UN1564
barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate
HQYOWPDUMCBSLQ-UHFFFAOYSA-L
barium(2+);2,3-dihydroxybutanedioate
6453753
98%
2.98

Chemical Identifiers

5908-81-6
285.397
HQYOWPDUMCBSLQ-UHFFFAOYSA-L
6453753
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2]
C4H4BaO6
MFCD00054437
barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate
barium(2+);2,3-dihydroxybutanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

EINECSNumber : 227-615-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only