CAS RN 5908-81-6
CAS RN 5908-81-6
IUPAC Name:
barium(2+) 2,3-dihydroxybutanedioate
Synonyms:
barium(2+) tartrate
Molecular Weight (g/mol):
285.4
Molecular Formula:
C4H4BaO6
InChi Key:
HQYOWPDUMCBSLQ-UHFFFAOYNA-L
SMILES:
[Ba++].OC(C(O)C([O-])=O)C([O-])=O
1
–
1
of
1
results
1
Barium tartrate, 98%
CAS: 5908-81-6 Molecular Formula: C4H4BaO6 Molecular Weight (g/mol): 285.397 MDL Number: MFCD00054437 InChI Key: HQYOWPDUMCBSLQ-UHFFFAOYSA-L Synonym: barium tartrate,barium 2+ tartrate,acmc-20alqr,barium 2+ ion tartrate,barium 2+ ; 2,3-dihydroxybutanedioate PubChem CID: 6453753 IUPAC Name: barium(2+);2,3-dihydroxybutanedioate SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2]