missing translation for 'onlineSavingsMsg'
Learn More

Barium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLBA22Y

Catalog No. CLBA2-2Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Barium Standard
7440-39-3 , 7732-18-5 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00134031
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ICP/ICP-MS Standard
[Ba]
barium
125 mL
CHEBI:32595
Triple-leached LDPE bottle
Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
0.1,2.0,97.9
Ba
ICP-MS
Standard Solution
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
2% HNO3
137.33
5355457
ICP/ICP-MS

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
CHEBI:32595
[Ba]
Ba
MFCD00134031
5355457
barium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only