missing translation for 'onlineSavingsMsg'
Learn More

Barium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLBA22T

 View more versions of this product

Catalog No. PLBA22T


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Barium Standard
7440-39-3 , 7732-18-5 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00134031
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
2% HNO3
137.33
5355457
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,2.0,97.9
Ba
AA, ICP
Certified Reference Material
[Ba]
barium
250 mL
CHEBI:32595
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLBA2-2Y, -2T, -2X    10,000    PLBA2-3Y, -3X
2-5% HCl    1000μg/mL    PLBA1-2X

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
CHEBI:32595
[Ba]
Ba
MFCD00134031
5355457
barium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only