missing translation for 'onlineSavingsMsg'
Learn More

Barium Standard For ICP, 1000μg/mL in 2% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLBA22X

Catalog No. PLBA2-2X

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Barium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00003448
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
AYJRCSIUFZENHW-UHFFFAOYSA-L
2% HNO3
197.34
10563
Triple-leached LDPE bottle
513-77-9,7732-18-5
CBaO3
AA, ICP
Certified Reference Material
[Ba++].[O-]C([O-])=O
barium(2+);carbonate
500 mL
Assurance

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+);carbonate
CBaO3
MFCD00003448
10563
[Ba++].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only