missing translation for 'onlineSavingsMsg'
Learn More

Barium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLBA23X

 View more versions of this product

Catalog No. PLBA2-3X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Barium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00134031
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
5% HNO3
137.33
5355457
Assurance
Triple-leached LDPE bottle
7440-39-3 , 7732-18-5
Ba
AA, ICP
Certified Reference Material
[Ba]
barium
500 mL
CHEBI:32595

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
CHEBI:32595
[Ba]
Ba
MFCD00134031
5355457
barium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause cancer. Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only